siRNA Details
Gene Symbol: EIF4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK310244.1 | - | 2 | 73 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK095174.1 | - | 1 | 1291 | Chr. 12 - 53007339 - 53009494 [+] on Build GRCh38 |
