siRNA Details
| Gene Symbol: MGC24103 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021795.1 | - | 1 | 91 | ||
| BC020879.1 | - | 1 | 119 | 4613905 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MGC24103| Gene Symbol: MGC24103 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021795.1 | - | 1 | 91 | ||
| BC020879.1 | - | 1 | 119 | 4613905 | ||