siRNA Details
| Gene Symbol: LINC01475 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120618.1 | 4 | 625 | |||
| GenBank mRNA | DQ372722.1 | - | 4 | 629 | ||
| DQ372723.1 | - | 2 | 196 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01475| Gene Symbol: LINC01475 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120618.1 | 4 | 625 | |||
| GenBank mRNA | DQ372722.1 | - | 4 | 629 | ||
| DQ372723.1 | - | 2 | 196 | |||