siRNA Details
| Gene Symbol: PRKCE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005264431.3 | XP_005264488.1 | 11 | 7191 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK023865.1 | - | 2 | 1017 | Chr. 2 - 46013055 - 46016909 [+] on Build GRCh38 | |

PRKCE
Genomic Map