siRNA Details
| Gene Symbol: MARC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317338.1 | NP_001304267.1 | 8 | 1692 | ||
| NM_017898.4 | NP_060368.2 | 8 | 1685 | |||
| XM_006711407.2 | XP_006711470.1 | 7 | 2241 | |||
| XM_011509684.1 | XP_011507986.1 | 8 | 1209 | |||
| XM_017001581.1 | XP_016857070.1 | 5 | 1377 | |||
| GenBank mRNA | AK000583.1 | - | 1 | 129 | ||
| BC010366.1 | - | 7 | 1020 | |||
| BC015829.2 | - | 6 | 955 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK125512.1 | - | 1 | 3066 | Chr. 1 - 220781213 - 220784803 [+] on Build GRCh38 | |

MARC2
Genomic Map