siRNA Details
| Gene Symbol: CXorf23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC093092.1 | - | 1 | 462 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CD102745.1 | - | 1 | 464 | Chr. X - 19965743 - 19966464 [-] on Build GRCh38 | |

CXorf23
Genomic Map