siRNA Details
| Gene Symbol: CRKL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA844269.1 | - | 1 | 43 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK124971.1 | - | 2 | 1972 | Chr. 22 - 20953534 - 20957814 [-] on Build GRCh38 | |

CRKL
Genomic Map