siRNA Details
| Gene Symbol: RNF40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK054773.1 | - | 1 | 125 | ||
| AK293710.1 | - | 9 | 1264 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RNF40| Gene Symbol: RNF40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK054773.1 | - | 1 | 125 | ||
| AK293710.1 | - | 9 | 1264 | |||