siRNA Details
| Gene Symbol: RBFADN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF070628.1 | - | 1 | 1434 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BM930419.1 | - | 1 | 289 | Chr. 18 - 80096763 - 80097092 [+] on Build GRCh38 | |

RBFADN
Genomic Map