siRNA Details
| Gene Symbol: ANKRD30BP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033891.1 | 5 | 821 | |||
| GenBank mRNA | AK098688.1 | - | 7 | 880 | ||
| BC031237.1 | - | 5 | 778 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ANKRD30BP3| Gene Symbol: ANKRD30BP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033891.1 | 5 | 821 | |||
| GenBank mRNA | AK098688.1 | - | 7 | 880 | ||
| BC031237.1 | - | 5 | 778 | |||