siRNA Details
| Gene Symbol: HOMER2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC044934.1 | - | 2 | 1222 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK128296.1 | - | 1 | 2073 | Chr. 15 - 82956793 - 82960241 [-] on Build GRCh38 | |

HOMER2
Genomic Map