siRNA Details
| Gene Symbol: SERF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199878.1 | NP_001186807.1 | 1 | 41 | ||
| GenBank mRNA | DA302996.1 | - | 1 | 41 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BU155533.1 | - | 53 | Chr. - - [-] on Build | ||

SERF2
Genomic Map