siRNA Details
| Gene Symbol: BACE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012105.4 | NP_036237.2 | 9 | 8321 | ||
| NM_138991.2 | NP_620476.1 | 8 | 8171 | |||
| NM_138992.2 | NP_620477.1 | 8 | 8152 | |||
| GenBank mRNA | AK023647.1 | - | 1 | 2827 | ||
| BC020582.1 | - | 1 | 2703 | |||

BACE2
Genomic Map