siRNA Details
| Gene Symbol: THTPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK090747.1 | - | 3 | 1359 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK126666.1 | - | 1 | 2550 | Chr. 14 - 23540563 - 23544283 [+] on Build GRCh38 | |

THTPA
Genomic Map