siRNA Details
| Gene Symbol: ZBED3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK098256.1 | - | 2 | 538 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DB253042.1 | - | 2 | 538 | Chr. 5 - 77139249 - 77146622 [+] on Build GRCh38 | |

ZBED3-AS1
Genomic Map