siRNA Details
| Gene Symbol: LOC101929261 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134277.1 | 2 | 620 | |||
| GenBank mRNA | BC030121.2 | - | 2 | 620 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG700797.1 | - | 2 | 625 | Chr. 5 - 7347044 - 7347977 [-] on Build GRCh38 | |

LOC101929261
Genomic Map