siRNA Details
| Gene Symbol: HSPH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286503.1 | NP_001273432.1 | 17 | 4431 | ||
| NM_001286504.1 | NP_001273433.1 | 18 | 4424 | |||
| NM_001286505.1 | NP_001273434.1 | 16 | 4190 | |||
| NM_006644.3 | NP_006635.2 | 18 | 4563 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CR611122.1 | - | 1 | 706 | Chr. 13 - 31135049 - 31136476 [-] on Build GRCh38 | |

HSPH1
Genomic Map