siRNA Details
| Gene Symbol: CEP152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC029603.1 | - | 1 | 84 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000560237.1 | - | 1 | 95 | Chr. 15 - 48810701 - 48811588 [+] on Build GRCh38 | |

CEP152
Genomic Map