siRNA Details
| Gene Symbol: LOC107987345 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK057112.1 | - | 1 | 859 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DN994925.1 | - | 1 | 317 | Chr. Y - 19043252 - 19043854 [-] on Build GRCh38 | |

LOC107987345
Genomic Map