siRNA Details
| Gene Symbol: LOC107987345 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017030091.1 | XP_016885580.1 | 4 | 1610 | ||
| GenBank mRNA | AK057112.1 | - | 1 | 2009 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC107987345| Gene Symbol: LOC107987345 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017030091.1 | XP_016885580.1 | 4 | 1610 | ||
| GenBank mRNA | AK057112.1 | - | 1 | 2009 | ||