siRNA Details
| Gene Symbol: KLHL36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303451.1 | NP_001290380.1 | 4 | 6161 | ||
| NM_024731.3 | NP_079007.2 | 5 | 6350 | |||
| XM_005256149.2 | XP_005256206.1 | 5 | 6564 | |||
| XM_017023674.1 | XP_016879163.1 | 5 | 6312 | |||
| XM_017023675.1 | XP_016879164.1 | 5 | 6332 | |||
| GenBank mRNA | AK056197.1 | - | 1 | 1344 | ||
| AK056428.1 | - | 1 | 3148 | |||
| AK074056.1 | - | 2 | 5277 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK057887.1 | - | 1184 | Chr. - - [-] on Build | ||

KLHL36
Genomic Map