siRNA Details
| Gene Symbol: LOC101928131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121677.1 | 1 | 41 | |||
| GenBank mRNA | BC039474.1 | - | 1 | 41 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BM450007.1 | - | 1 | 40 | Chr. 4 - 165731364 - 165743070 [-] on Build GRCh38 | |

LOC101928131
Genomic Map