siRNA Details
Gene Symbol: LINC01505 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104145.1 | 1 | 121 | |||
GenBank mRNA | AK093363.1 | - | 1 | 121 | ||
BC034311.1 | - | 1 | 121 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01505 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104145.1 | 1 | 121 | |||
GenBank mRNA | AK093363.1 | - | 1 | 121 | ||
BC034311.1 | - | 1 | 121 |