siRNA Details
| Gene Symbol: KIRREL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286349.1 | NP_001273278.1 | 13 | 5199 | ||
| NM_018240.6 | NP_060710.3 | 15 | 5499 | |||
| XM_005245305.4 | XP_005245362.1 | 15 | 5204 | |||
| GenBank mRNA | AK090554.1 | - | 1 | 1882 | ||
| AK094163.1 | - | 1 | 34 | |||
| AK096361.1 | - | 1 | 1118 | |||
| AK098412.1 | - | 1 | 624 | |||

KIRREL
Genomic Map