siRNA Details
| Gene Symbol: LINC00970 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104091.1 | 5 | 550 | |||
| GenBank mRNA | BC041008.1 | - | 5 | 555 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI544897.1 | - | 5 | 555 | Chr. 1 - 168904133 - 169087007 [-] on Build GRCh38 | |

LINC00970
Genomic Map