siRNA Details
| Gene Symbol: AJAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011541786.2 | XP_011540088.1 | 7 | 8770 | ||
| XM_011541787.2 | XP_011540089.1 | 6 | 8438 | |||
| GenBank mRNA | CR936609.1 | - | 1 | 2390 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AJAP1| Gene Symbol: AJAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011541786.2 | XP_011540088.1 | 7 | 8770 | ||
| XM_011541787.2 | XP_011540089.1 | 6 | 8438 | |||
| GenBank mRNA | CR936609.1 | - | 1 | 2390 | ||