siRNA Details
| Gene Symbol: NCAPH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281710.1 | NP_001268639.1 | 18 | 4645 | ||
| NM_001281711.1 | NP_001268640.1 | 18 | 4606 | |||
| NM_001281712.1 | NP_001268641.1 | 17 | 4425 | |||
| NM_015341.4 | NP_056156.2 | 18 | 4678 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CR608905.1 | - | 1 | 1051 | Chr. 2 - 96374664 - 96377067 [+] on Build GRCh38 | |

NCAPH
Genomic Map