siRNA Details
| Gene Symbol: NCAPH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281710.1 | NP_001268639.1 | 18 | 5116 | ||
| NM_001281711.1 | NP_001268640.1 | 18 | 5077 | |||
| NM_001281712.1 | NP_001268641.1 | 17 | 4896 | |||
| NM_015341.4 | NP_056156.2 | 18 | 5149 | |||

NCAPH
Genomic Map