siRNA Details
| Gene Symbol: AGAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037131.2 | NP_001032208.1 | 18 | 10746 | ||
| NM_014914.4 | NP_055729.2 | 17 | 10587 | |||
| XM_005246059.4 | XP_005246116.1 | 18 | 10052 | |||
| XM_006712234.3 | XP_006712297.1 | 20 | 10287 | |||
| XM_006712235.3 | XP_006712298.1 | 19 | 10411 | |||
| XM_006712237.3 | XP_006712300.2 | 19 | 11068 | |||
| XM_006712239.3 | XP_006712302.2 | 20 | 11083 | |||
| XM_011510547.2 | XP_011508849.1 | 19 | 11004 | |||
| XM_011510548.2 | XP_011508850.1 | 19 | 10922 | |||
| XM_011510549.2 | XP_011508851.1 | 18 | 10885 | |||
| XM_017003282.1 | XP_016858771.1 | 19 | 10957 | |||

AGAP1
Genomic Map