siRNA Details
| Gene Symbol: LRCH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005269365.2 | XP_005269422.1 | 20 | 10364 | ||
| XM_005269367.2 | XP_005269424.1 | 19 | 10292 | |||
| XM_006713791.3 | XP_006713854.1 | 21 | 7443 | |||
| XM_006713792.3 | XP_006713855.1 | 20 | 7371 | |||
| XM_006713793.2 | XP_006713856.1 | 20 | 7335 | |||
| XM_017007352.1 | XP_016862841.1 | 18 | 10238 | |||
| GenBank mRNA | AK125235.1 | - | 1 | 1894 | ||
| BC008625.1 | - | 1 | 1302 | |||

LRCH3
Genomic Map