siRNA Details
Gene Symbol: IPO5P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103741.1 | 3 | 289 | |||
NR_103742.1 | 2 | 214 | ||||
GenBank mRNA | AK023040.1 | - | 3 | 268 | ||
AK056739.1 | - | 1 | 928 | |||
AK056856.1 | - | 1 | 2170 | |||
DA183233.1 | - | 2 | 179 | |||
DA185451.1 | - | 3 | 274 | |||
DA510220.1 | - | 3 | 296 | |||
DA723810.1 | - | 2 | 214 |
