siRNA Details
Gene Symbol: IPO5P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103741.1 | 3 | 310 | |||
NR_103742.1 | 2 | 235 | ||||
GenBank mRNA | AK056739.1 | - | 1 | 949 | ||
AK056856.1 | - | 1 | 2191 | |||
DA183233.1 | - | 2 | 200 | |||
DA185451.1 | - | 3 | 295 | |||
DA510220.1 | - | 3 | 317 | |||
DA723810.1 | - | 2 | 235 |
