siRNA Details
| Gene Symbol: LOC101929457 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135692.1 | 1 | 685 | |||
| GenBank mRNA | BC062542.1 | - | 1 | 685 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA726967.1 | - | 1 | 354 | Chr. 15 - 77525926 - 77531240 [+] on Build GRCh38 | |

LOC101929457
Genomic Map