siRNA Details
| Gene Symbol: LOC101929457 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135692.1 | 2 | 3685 | |||
| GenBank mRNA | BC062542.1 | - | 2 | 3685 | ||
| BF516416.1 | - | 1 | 219 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929457| Gene Symbol: LOC101929457 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135692.1 | 2 | 3685 | |||
| GenBank mRNA | BC062542.1 | - | 2 | 3685 | ||
| BF516416.1 | - | 1 | 219 | |||