siRNA Details
Gene Symbol: P2RX7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011538419.2 | XP_011536721.1 | 12 | 6261 | ||
XM_011538420.2 | XP_011536722.1 | 11 | 5928 | |||
XM_017019364.1 | XP_016874853.1 | 13 | 6574 | |||
XM_017019365.1 | XP_016874854.1 | 12 | 6302 | |||
XM_017019366.1 | XP_016874855.1 | 12 | 6289 | |||
XM_017019367.1 | XP_016874856.1 | 11 | 6127 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK123448.1 | - | 2 | 1434 | Chr. 12 - 121187536 - 121189446 [+] on Build GRCh38 |
