siRNA Details
| Gene Symbol: P2RX7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011538419.2 | XP_011536721.1 | 12 | 5544 | ||
| XM_011538420.2 | XP_011536722.1 | 11 | 5211 | |||
| XM_017019364.1 | XP_016874853.1 | 13 | 5857 | |||
| XM_017019365.1 | XP_016874854.1 | 12 | 5585 | |||
| XM_017019366.1 | XP_016874855.1 | 12 | 5572 | |||
| XM_017019367.1 | XP_016874856.1 | 11 | 5410 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK123448.1 | - | 1 | 1039 | Chr. 12 - 121187536 - 121189446 [+] on Build GRCh38 | |

P2RX7
Genomic Map