siRNA Details
| Gene Symbol: LOC100130976 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK128633.1 | - | 1 | 1731 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100130976| Gene Symbol: LOC100130976 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK128633.1 | - | 1 | 1731 | ||