siRNA Details
| Gene Symbol: LOC101927849 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125887.1 | 7 | 1675 | |||
| GenBank mRNA | BC040325.1 | - | 7 | 1675 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927849| Gene Symbol: LOC101927849 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125887.1 | 7 | 1675 | |||
| GenBank mRNA | BC040325.1 | - | 7 | 1675 | ||