siRNA Details
| Gene Symbol: LINC00423 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047020.1 | 4, 5 | 898 | |||
| GenBank mRNA | BC024029.1 | - | 4, 5 | 903 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00423| Gene Symbol: LINC00423 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047020.1 | 4, 5 | 898 | |||
| GenBank mRNA | BC024029.1 | - | 4, 5 | 903 | ||