siRNA Details
Gene Symbol: LINC01166 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120626.1 | 2 | 2726 | |||
NR_120627.1 | 2 | 2560 | ||||
GenBank mRNA | AK095503.1 | - | 2 | 2560 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01166 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120626.1 | 2 | 2726 | |||
NR_120627.1 | 2 | 2560 | ||||
GenBank mRNA | AK095503.1 | - | 2 | 2560 |