siRNA Details
| Gene Symbol: LINC01166 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120627.1 | 1 | 88 | |||
| GenBank mRNA | AK095503.1 | - | 1 | 88 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA483121.1 | - | 1 | 87 | Chr. 10 - 132945818 - 132965422 [-] on Build GRCh38 | |

LINC01166
Genomic Map