siRNA Details
| Gene Symbol: CHAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017022563.1 | XP_016878052.1 | 1 | 1139 | ||
| XM_017022565.1 | XP_016878054.1 | 1 | 1139 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK094272.1 | - | 1 | 814 | Chr. 21 - 21080468 - 21082281 [+] on Build GRCh38 | |

CHAC1
Genomic Map