siRNA Details
| Gene Symbol: EIF2AK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011532987.2 | XP_011531289.1 | 17 | 9876 | ||
| XM_017004503.1 | XP_016859992.1 | 16 | 9743 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

EIF2AK2| Gene Symbol: EIF2AK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011532987.2 | XP_011531289.1 | 17 | 9876 | ||
| XM_017004503.1 | XP_016859992.1 | 16 | 9743 | |||