siRNA Details
| Gene Symbol: SLC35F5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005263799.3 | XP_005263856.1 | 16 | 8724 | ||
| XM_011511921.2 | XP_011510223.1 | 17 | 8175 | |||
| XM_011511922.2 | XP_011510224.1 | 17 | 8392 | |||
| XM_011511923.2 | XP_011510225.1 | 16 | 8077 | |||
| XM_017005028.1 | XP_016860517.1 | 15 | 8038 | |||

SLC35F5
Genomic Map