siRNA Details
| Gene Symbol: CHCHD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304354.1 | NP_001291283.1 | 1 | 17 | ||
| GenBank mRNA | BM563093.1 | - | 1 | 17 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK024631.1 | - | 1 | 442 | Chr. 2 - 112584449 - 112585920 [+] on Build GRCh38 | |

CHCHD5
Genomic Map