siRNA Details
| Gene Symbol: HOXB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017024563.1 | XP_016880052.1 | 1 | 1373 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BF799301.1 | - | 1 | 226 | Chr. 17 - 48583419 - 48583736 [-] on Build GRCh38 | |

HOXB3
Genomic Map