siRNA Details
| Gene Symbol: B3GNT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011513225.2 | XP_011511527.1 | 1 | 576 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK126337.1 | - | 1 | 576 | Chr. 3 - 183265257 - 183269014 [+] on Build GRCh38 | |

B3GNT5
Genomic Map