siRNA Details
| Gene Symbol: LOC101929745 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134272.1 | 2 | 2137 | |||
| GenBank mRNA | BC028978.1 | - | 2 | 2137 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929745| Gene Symbol: LOC101929745 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134272.1 | 2 | 2137 | |||
| GenBank mRNA | BC028978.1 | - | 2 | 2137 | ||