siRNA Details
| Gene Symbol: ALG12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BM145575.1 | - | 1 | 133 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK126604.1 | - | 2 | 954 | Chr. 22 - 49902228 - 49904576 [+] on Build GRCh38 | |

ALG12
Genomic Map