siRNA Details
| Gene Symbol: LOC101928517 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110732.1 | 3 | 652 | |||
| GenBank mRNA | BC045766.1 | - | 3 | 652 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000600074.1 | - | 3 | 652 | Chr. 19 - 51152923 - 51181966 [-] on Build GRCh38 | |

LOC101928517
Genomic Map